Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042368.2 | NP_001035827.1 | 6 | 902 | ||
NM_001271774.1 | NP_001258703.1 | 6 | 814 | |||
NM_001271775.1 | NP_001258704.1 | 6 | 907 | |||
NM_001271776.1 | NP_001258705.1 | 6 | 887 | |||
NM_006266.3 | NP_006257.1 | 6 | 923 | |||
GenBank mRNA | AB037729.1 | - | 2 | 215 | ||
AF027169.1 | - | 6 | 818 | |||
AF295773.1 | - | 6 | 838 | |||
AK056462.1 | - | 5 | 600 | |||
AK074114.1 | - | 6 | 793 | |||
AK090450.1 | - | 2 | 249 | |||
AK127524.1 | - | 6 | 907 | |||
AK301470.1 | - | 6 | 774 | |||
BC059362.1 | - | 6 | 877 | 30340922 |