Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4921509C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198655.2 | NP_941057.1 | 2 | 282 | ||
GenBank mRNA | AK029504.1 | - | 2 | 284 | ||
BC119071.1 | - | 1 | 112 | 8733820 | ||
BC119073.1 | - | 1 | 112 | 8733822 |
Gene Symbol: Gm14151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097977.1 | NP_001091446.1 | 2 | 282 |