Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stpg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198659.2 | NP_941061.1 | 6 | 717 | ||
GenBank mRNA | AK046955.1 | - | 6 | 718 | ||
AK161389.1 | - | 6 | 712 | |||
BC138108.1 | - | 6 | 756 | 8861127 | ||
BC138109.1 | - | 6 | 756 | 8861128 |