Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC061212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198667.1 | NP_941069.1 | 3 | 517 | ||
GenBank mRNA | BC061212.1 | - | 3 | 517 | 30250942 |
Gene Symbol: A430089I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177913.4 | NP_808581.1 | 3 | 517 | ||
GenBank mRNA | AK040373.1 | - | 3 | 518 | ||
BC147369.1 | - | 3 | 484 | 9056433 |
Gene Symbol: BC080696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC080696.1 | - | 2 | 442 |
Gene Symbol: Gm7978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270457.1 | NP_001257386.1 | 3 | 517 |
Gene Symbol: LOC545763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN677185.1 | - | 2 | 412 |