Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270494.1 | NP_001257423.1 | 2 | 156 | ||
GenBank mRNA | BC147388.1 | - | 2 | 156 | 9056468 | |
BC147389.1 | - | 2 | 156 | 9056459 |
Gene Symbol: Gm10662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201364.1 | NP_001188293.1 | 4 | 485 |
Gene Symbol: Gm4567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037248.3 | NP_001032325.1 | 3 | 336 | ||
GenBank mRNA | AK139578.1 | - | 3 | 337 |
Gene Symbol: Gm5891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034904.2 | NP_001030076.1 | 4 | 487 | ||
GenBank mRNA | AK139650.1 | - | 4 | 487 | ||
AK163219.1 | - | 358 |