Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030981.2 | NP_112243.1 | 5 | 468 | ||
XM_017018378.1 | XP_016873867.1 | 5 | 527 | |||
GenBank mRNA | AF092437.1 | - | 5 | 386 | ||
AJ245875.1 | - | 4 | 375 | |||
AK292086.1 | - | 5 | 436 | |||
AK315333.1 | - | 5 | 429 | |||
AL136635.1 | - | 5 | 420 | |||
BC028331.1 | - | 5 | 419 | |||
BC071169.1 | - | 5 | 407 | 6026121 | ||
CR533462.1 | - | 4 | 373 |