Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029876.1 | NP_001025047.1 | 3 | 2227 | ||
GenBank mRNA | AK122209.1 | - | 1 | 1864 | ||
AK133906.1 | - | 4 | 2541 | |||
BC096020.1 | - | 1 | 611 | |||
BC151125.1 | - | 3 | 2306 | 9088038 |