Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164681.1 | NP_001158153.1 | 8 | 965 | ||
NM_198672.1 | NP_941074.1 | 9 | 1106 | |||
GenBank mRNA | AK138932.1 | - | 9 | 1105 | ||
AK149499.1 | - | 5 | 526 | |||
BC035526.1 | - | 8 | 961 | |||
BC048380.1 | - | 9 | 1087 | |||
BC057187.1 | - | 9 | 1091 | 5098677 | ||
BC057188.1 | - | 9 | 1091 | 5100121 | ||
BC061004.1 | - | 8 | 963 | 30315941 | ||
L11333.1 | - | 9 | 1081 | |||
S64130.1 | - | 9 | 1082 |