Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004062.2 | NP_001004062.1 | 5, 6 | 559 | ||
GenBank mRNA | AK129173.1 | - | 5, 6 | 556 | ||
BC056137.1 | - | 5, 6 | 565 | |||
BC068139.1 | - | 5, 6 | 565 | |||
BC080308.1 | - | 5, 6 | 559 | 5719562 |