Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004062.2 | NP_001004062.1 | 7, 8 | 688 | ||
GenBank mRNA | AK129173.1 | - | 7, 8 | 685 | ||
BC056137.1 | - | 7, 8 | 694 | |||
BC068139.1 | - | 7, 8 | 694 | |||
BC080308.1 | - | 7, 8 | 688 | 5719562 |