Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201372.3 | NP_958760.2 | 6 | 580 | ||
GenBank mRNA | AK052757.1 | - | 6 | 623 | ||
AK162606.1 | - | 5 | 882 | |||
AK171754.1 | - | 856 | ||||
BC055929.1 | - | 6 | 580 | |||
BC096560.1 | - | 6 | 585 | 1527981 | ||
BC116877.1 | - | 6 | 622 | 40126196 | ||
BC116879.1 | - | 6 | 622 | 40126198 | ||
BC144712.1 | - | 6 | 622 | |||
BC144713.1 | - | 6 | 622 |