Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201372.3 | NP_958760.2 | 7 | 661 | ||
GenBank mRNA | AK052757.1 | - | 7 | 704 | ||
AK162606.1 | - | 5 | 1077 | |||
AK171754.1 | - | 1051 | ||||
BC055929.1 | - | 7 | 661 | |||
BC096560.1 | - | 7 | 666 | 1527981 |