Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brwd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081477.1 | NP_001074946.1 | 5 | 442 | ||
GenBank mRNA | AK042444.1 | - | 434 | |||
AK050539.1 | - | 5 | 428 | |||
AK163354.1 | - | 5 | 434 |