Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002890.2 | NP_002881.1 | 17 | 2503 | ||
NM_022650.2 | NP_072179.1 | 17 | 1897 | |||
XM_011543525.1 | XP_011541827.1 | 17 | 2955 | |||
GenBank mRNA | AB209106.1 | - | 17 | 2503 | ||
AK300263.1 | - | 18 | 2004 | |||
AK300269.1 | - | 17 | 1830 | |||
AK300406.1 | - | 18 | 1923 | |||
AK312739.1 | - | 17 | 1882 | |||
BC033015.2 | - | 17 | 2491 | 4829173 | ||
BC107870.1 | - | 17 | 1883 | |||
CR749722.1 | - | 18 | 3013 | |||
M23379.1 | - | 17 | 2422 | |||
M23612.1 | - | 17 | 1872 |