Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303245.1 | NP_001290174.1 | 23 | 2435 | ||
NM_001303246.1 | NP_001290175.1 | 24 | 2444 | |||
NM_006506.3 | NP_006497.2 | 23 | 2432 | |||
XM_011513059.2 | XP_011511361.1 | 24 | 3584 | |||
XM_017006968.1 | XP_016862457.1 | 24 | 2568 | |||
XM_017006969.1 | XP_016862458.1 | 24 | 2564 | |||
XM_017006972.1 | XP_016862461.1 | 22 | 2299 | |||
GenBank mRNA | AB209161.1 | - | 8 | 771 | ||
AF115573.1 | - | 23 | 2395 | |||
AK292016.1 | - | 23 | 2433 | |||
D78155.1 | - | 23 | 2398 | |||
D82880.1 | - | 24 | 2407 |