Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm1322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033477.3 | NP_001028649.2 | 2 | 594 | ||
GenBank mRNA | AK158464.1 | - | 2 | 595 |
Gene Symbol: Xirp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KM273012.1 | - | 404 |