Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135255.1 | NP_001128727.1 | 4 | 574 | ||
NM_001135256.1 | NP_001128728.1 | 4 | 477 | |||
NM_005610.2 | NP_005601.1 | 4 | 577 | |||
GenBank mRNA | AK222779.1 | - | 4 | 480 | ||
AK299251.1 | - | 4 | 477 | |||
AK312571.1 | - | 4 | 515 | |||
BC003092.1 | - | 4 | 504 | 3356558 | ||
BC015123.1 | - | 2 | 238 | |||
BC053904.1 | - | 4 | 490 | 6526200 | ||
BC075836.1 | - | 4 | 445 | 6338602 | ||
BT007309.1 | - | 4 | 418 | |||
X71810.1 | - | 4 | 491 | |||
X74262.1 | - | 4 | 502 |