Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323281.1 | NP_001310210.1 | 11 | 1895 | ||
NM_001323282.1 | NP_001310211.1 | 12 | 1960 | |||
NM_002895.4 | NP_002886.2 | 12 | 1822 | |||
NM_183404.3 | NP_899662.1 | 12 | 1822 | |||
XM_006723842.2 | XP_006723905.1 | 11 | 1533 | |||
XM_011528955.2 | XP_011527257.1 | 12 | 1598 | |||
XM_011528958.2 | XP_011527260.1 | 12 | 1598 | |||
XM_017027992.1 | XP_016883481.1 | 13 | 1778 | |||
GenBank mRNA | AK290380.1 | - | 12 | 1598 | ||
AK308096.1 | - | 12 | 1583 | |||
BC032247.1 | - | 12 | 1605 | 4521266 | ||
L14812.1 | - | 12 | 1603 | |||
M74547.1 | - | 10 | 1136 |