Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323608.1 | NP_001310537.1 | 9 | 1350 | ||
NM_001323609.1 | NP_001310538.1 | 9 | 1350 | |||
NM_001323610.1 | NP_001310539.1 | 9 | 1350 | |||
NM_001323611.1 | NP_001310540.1 | 9 | 1099 | |||
NM_005611.3 | NP_005602.3 | 9 | 1331 | |||
XM_011523253.2 | XP_011521555.1 | 7 | 724 | |||
XM_017023513.1 | XP_016879002.1 | 6 | 888 | |||
GenBank mRNA | AK300047.1 | - | 9 | 1069 | ||
AK304283.1 | - | 6 | 798 | |||
AK307881.1 | - | 9 | 1308 | |||
BC034490.1 | - | 9 | 1321 | 4838444 | ||
BX537767.1 | - | 8 | 912 | |||
S67171.1 | - | 9 | 1241 | |||
X74594.1 | - | 9 | 1042 | |||
X76061.1 | - | 9 | 1282 |