Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198864.2 | NP_942564.2 | 2 | 2122 | ||
GenBank mRNA | AK139378.1 | - | 2 | 2245 | ||
AK158856.1 | - | 2 | 1977 | |||
BC058541.1 | - | 1 | 511 | |||
BC066059.1 | - | 2 | 2122 | 6844461 |