Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002898.3 | NP_002889.1 | 7 | 889 | ||
XM_005269059.4 | XP_005269116.1 | 7 | 757 | |||
XM_005269060.4 | XP_005269117.1 | 7 | 757 | |||
XM_005269061.3 | XP_005269118.1 | 7 | 757 | |||
XM_006719541.3 | XP_006719604.1 | 7 | 757 | |||
XM_006719542.3 | XP_006719605.1 | 7 | 757 | |||
XM_006719543.3 | XP_006719606.1 | 7 | 757 | |||
XM_006719544.3 | XP_006719607.1 | 7 | 757 | |||
XM_011538637.2 | XP_011536939.1 | 7 | 701 | |||
XM_011538638.2 | XP_011536940.1 | 8 | 1023 | |||
XM_011538639.2 | XP_011536941.1 | 7 | 757 | |||
XM_011538640.2 | XP_011536942.1 | 7 | 757 | |||
XM_011538642.2 | XP_011536944.1 | 5 | 487 | |||
XM_017019775.1 | XP_016875264.1 | 7 | 696 | |||
XM_017019776.1 | XP_016875265.1 | 3 | 181 | |||
XM_017019777.1 | XP_016875266.1 | 6 | 605 | |||
XM_017019778.1 | XP_016875267.1 | 3 | 197 | |||
GenBank mRNA | AK293315.1 | - | 6 | 489 | ||
AK304540.1 | - | 6 | 549 | |||
AK308885.1 | - | 5 | 585 | |||
BC027863.1 | - | 7 | 698 | 5202634 | ||
BC072679.1 | - | 7 | 762 | 6048623 | ||
D28483.1 | - | 7 | 706 |