Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021562.1 | NP_067537.1 | 1 | 753 | ||
GenBank mRNA | AF247735.1 | - | 1 | 660 | ||
BC145965.1 | - | 1 | 776 | 40131268 | ||
BC145967.1 | - | 1 | 776 | 40131270 |