Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tigd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207278.2 | NP_997161.1 | 1 | 708 | ||
GenBank mRNA | AK081667.1 | - | 1 | 710 |