Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323517.1 | NP_001310446.1 | 6 | 785 | ||
NM_001323518.1 | NP_001310447.1 | 6 | 637 | |||
NM_006744.3 | NP_006735.2 | 6 | 699 | |||
GenBank mRNA | BC020633.1 | - | 6 | 699 | 4767481 | |
BX495987.1 | - | 2 | 259 | |||
X00129.1 | - | 6 | 660 |