Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323517.1 | NP_001310446.1 | 5 | 675 | ||
NM_001323518.1 | NP_001310447.1 | 5 | 527 | |||
NM_006744.3 | NP_006735.2 | 5 | 589 | |||
GenBank mRNA | BC020633.1 | - | 5 | 589 | 4767481 | |
BX495987.1 | - | 1 | 149 | |||
X00129.1 | - | 5 | 550 |