Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003667.1 | NP_001003667.1 | 7 | 1230 | ||
GenBank mRNA | AK132513.1 | - | 7 | 1318 | ||
BC107395.2 | - | 7 | 1230 | 40058152 | ||
BC107396.2 | - | 7 | 1230 | 40058154 | ||
BC141259.1 | - | 7 | 1253 | 9055871 |