Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039666.1 | NP_001034755.1 | 5 | 915 | ||
GenBank mRNA | BC107359.1 | - | 5 | 915 | ||
BC107360.2 | - | 5 | 913 | |||
BC119521.2 | - | 3 | 675 | 40055957 | ||
BC119522.2 | - | 5 | 915 | 40055959 |