Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080924.2 | NP_001074393.1 | 2 | 480 | ||
NM_001290501.1 | NP_001277430.1 | 3 | 327 | |||
GenBank mRNA | AK133342.1 | - | 3 | 328 | ||
AK163639.1 | - | 2 | 480 | |||
BC151080.1 | - | 3 | 201 | 9087993 | ||
BC151083.1 | - | 3 | 201 | 9087996 |