Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001182.3 | NP_001001182.2 | 21 | 3367 | ||
GenBank mRNA | AK029231.1 | - | 15 | 2415 | ||
AK173180.1 | - | 21 | 3399 | |||
BC150814.1 | - | 23 | 3376 | 9087725 |