Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001495.2 | NP_001001495.2 | 3 | 371 | ||
GenBank mRNA | AK033534.1 | - | 3 | 175 | ||
BC104392.1 | - | 3 | 174 | |||
BC104393.1 | - | 3 | 174 | 40050409 | ||
EF585494.1 | - | 3 | 353 | |||
EF585495.1 | - | 3 | 353 |