Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002909.4 | NP_002900.2 | 6 | 724 | ||
GenBank mRNA | AK291981.1 | - | 6 | 692 | ||
BC005350.2 | - | 5 | 668 | 3950349 | ||
M18963.1 | - | 5 | 676 | |||
M27190.1 | - | 5 | 703 |