Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002909.4 | NP_002900.2 | 5, 6 | 541 | ||
GenBank mRNA | AF172331.1 | - | 4, 5 | 488 | ||
AK291981.1 | - | 5, 6 | 509 | |||
BC005350.2 | - | 4, 5 | 485 | 3950349 | ||
BM769434.1 | - | 5, 6 | 553 | |||
M18963.1 | - | 4, 5 | 493 | |||
M27190.1 | - | 4, 5 | 520 |