Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000475.4 | NP_000466.2 | 2 | 1304 | ||
GenBank mRNA | BC011564.1 | - | 2 | 1301 | 4637694 | |
HQ692832.1 | - | 2 | 1289 | |||
S74720.1 | - | 2 | 1523 |