Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163154.1 | NP_001156626.1 | 8 | 961 | ||
NM_007960.4 | NP_031986.1 | 11 | 1301 | |||
GenBank mRNA | AK032224.1 | - | 8 | 963 | ||
AK035914.1 | - | 9 | 1692 | |||
AK144713.1 | - | 11 | 1250 | |||
BC005645.1 | - | 11 | 1229 | 3257346 | ||
BE859344.1 | - | 1 | 76 | |||
CK619331.1 | - | 4 | 375 | |||
L10426.1 | - | 11 | 1250 |