Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027976.2 | NP_082252.1 | 14, 15 | 2145 | ||
GenBank mRNA | AK006541.1 | - | 3, 4 | 622 | ||
AK034942.1 | - | 2145 | ||||
AK145894.1 | - | 14, 15 | 1806 | |||
AK150542.1 | - | 14, 15 | 1811 | |||
AK150663.1 | - | 14, 15 | 1811 | |||
AK151378.1 | - | 14, 15 | 1600 | |||
AK161024.1 | - | 14, 15 | 1498 | |||
AK168019.1 | - | 2, 3 | 146 | |||
BC031544.1 | - | 14, 15 | 1579 | 3987201 |