Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278791.1 | NP_001265720.1 | 6 | 682 | ||
NM_001278792.1 | NP_001265721.1 | 5 | 473 | |||
NM_001278793.1 | NP_001265722.1 | 5 | 580 | |||
NM_002914.4 | NP_002905.2 | 5 | 478 | |||
NM_181471.2 | NP_852136.1 | 6 | 580 | |||
XM_006716080.2 | XP_006716143.1 | 4 | 302 | |||
XM_017012492.1 | XP_016867981.1 | 6 | 558 | |||
XM_017012493.1 | XP_016867982.1 | 4 | 326 | |||
GenBank mRNA | AB451243.1 | - | 6 | 505 | ||
AB451368.1 | - | 6 | 505 | |||
BC002813.1 | - | 5 | 408 | 3637774 | ||
BM563468.1 | - | 6 | 611 | |||
BM810551.1 | - | 5 | 385 | |||
BQ070526.1 | - | 6 | 530 | |||
BQ674131.1 | - | 5 | 431 | |||
BT007058.1 | - | 5 | 403 | |||
M87338.1 | - | 712 |