Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syndig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085521.2 | NP_001078990.1 | 2 | 440 | ||
GenBank mRNA | BB653505.1 | - | 2 | 440 | ||
BC147352.1 | - | 1 | 241 | 9056389 | ||
BC147353.1 | - | 1 | 241 | 9056409 | ||
BC147660.1 | - | 1 | 241 | 9087449 | ||
BC147661.1 | - | 1 | 241 | 9087450 | ||
CJ061528.1 | - | 2 | 429 |