Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Minos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163006.2 | NP_001156478.1 | 4 | 546 | ||
GenBank mRNA | AK004466.1 | - | 4 | 438 | ||
AK008362.1 | - | 4 | 430 | |||
AK008449.1 | - | 4 | 430 | |||
AK009189.1 | - | 4 | 429 | |||
AK009528.1 | - | 4 | 472 | |||
AK011553.1 | - | 4 | 429 | |||
AK030343.1 | - | 4 | 499 | |||
AK152509.1 | - | 4 | 472 | |||
AK153063.1 | - | 4 | 471 | |||
AK159039.1 | - | 4 | 425 | |||
AK159100.1 | - | 4 | 472 | |||
AK167818.1 | - | 4 | 471 | |||
BC055791.1 | - | 4 | 463 | 6410794 |