Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122678.2 | NP_001116150.1 | 4 | 1668 | ||
NM_001167792.1 | NP_001161264.1 | 3 | 1077 | |||
NM_001167793.1 | NP_001161265.1 | 4 | 1708 | |||
GenBank mRNA | AK145737.1 | - | 6 | 1964 |
Gene Symbol: D5Ertd577e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177187.4 | NP_796161.3 | 4 | 1670 | ||
GenBank mRNA | AK054417.1 | - | 4 | 1670 | ||
BC053383.1 | - | 3 | 1579 | |||
BC094905.1 | - | 3 | 1567 | 30047197 | ||
BC099941.1 | - | 4 | 1654 | 30031929 | ||
BC145804.1 | - | 4 | 1616 | 40131107 |
Gene Symbol: E330014E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122668.1 | NP_001116140.1 | 4 | 1790 | ||
GenBank mRNA | AK087741.1 | - | 4 | 1792 | ||
BB709961.1 | - | 1 | 85 |
Gene Symbol: Gm16367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031622.2 | NP_001026793.2 | 4 | 1695 | ||
GenBank mRNA | AK145705.1 | - | 4 | 1708 | ||
BC099967.1 | - | 4 | 1695 | 30027969 |
Gene Symbol: Gm16513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200055.1 | NP_001186984.1 | 1706 |
Gene Symbol: Gm3139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243937.1 | NP_001230866.1 | 4 | 1726 | ||
NM_001243938.1 | NP_001230867.1 | 4 | 1686 |