Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081265.1 | NP_001074734.1 | 9 | 1936 | ||
GenBank mRNA | BC053401.1 | - | 2 | 284 | ||
BC158066.1 | - | 9 | 1394 | 9007365 |