Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081265.1 | NP_001074734.1 | 10 | 1975 | ||
GenBank mRNA | BC053401.1 | - | 3 | 323 | ||
BC147492.1 | - | 10 | 1969 | 9056289 | ||
BC158066.1 | - | 10 | 1433 | 9007365 |