Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002918.4 | NP_002909.4 | 2, 3 | 583 | ||
XM_011528165.2 | XP_011526467.1 | 2, 3 | 583 | |||
XM_011528166.2 | XP_011526468.1 | 2, 3 | 519 | |||
XM_011528167.2 | XP_011526469.1 | 2, 3 | 1128 | |||
XM_011528168.2 | XP_011526470.1 | 2, 3 | 583 | |||
GenBank mRNA | A20498.1 | - | 2, 3 | 404 | ||
AK309824.1 | - | 2, 3 | 582 | |||
BC049826.1 | - | 2, 3 | 544 | 6043247 | ||
X58964.1 | - | 2, 3 | 404 |