Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000635.3 | NP_000626.2 | 7 | 883 | ||
NM_134433.2 | NP_602309.1 | 6 | 808 | |||
XM_011528171.2 | XP_011526473.1 | 8 | 849 | |||
XM_011528172.1 | XP_011526474.1 | 7 | 706 | |||
XM_017027107.1 | XP_016882596.1 | 6 | 733 | |||
XM_017027108.1 | XP_016882597.1 | 5 | 658 | |||
GenBank mRNA | AK024288.1 | - | 4 | 422 | ||
AK093977.1 | - | 2 | 1791 | |||
AK291196.1 | - | 6 | 687 | |||
BC028579.1 | - | 7 | 795 | 4829582 | ||
BC071571.1 | - | 7 | 764 | 5276369 | ||
BX648130.1 | - | 7 | 718 | |||
X76091.1 | - | 7 | 773 |