Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whamm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004185.3 | NP_001004185.3 | 10 | 2212 | ||
GenBank mRNA | AK220273.1 | - | 10 | 2212 | ||
BC042749.2 | - | 11 | 2047 | 5322109 | ||
BC050804.1 | - | 11 | 1381 | 6771347 |