Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079932.3 | NP_001073401.1 | 3 | 746 | ||
GenBank mRNA | AB231474.1 | - | 3 | 650 | ||
BC067209.1 | - | 3 | 312 | |||
BC147789.1 | - | 3 | 746 | 9087507 | ||
BC158111.1 | - | 3 | 731 | 9088185 | ||
BC158120.1 | - | 3 | 731 | 9088192 | ||
BC172156.1 | - | 3 | 746 |