Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170669.2 | NP_733769.1 | 2 | 178 | ||
GenBank mRNA | AK008350.1 | - | 3 | 189 | ||
AK010652.1 | - | 3 | 219 | |||
AK011514.1 | - | 3 | 192 | |||
AK012485.1 | - | 3 | 196 | |||
AK012618.1 | - | 3 | 192 | |||
AK019235.1 | - | 3 | 192 | |||
AK028135.1 | - | 3 | 192 | |||
AK032522.1 | - | 3 | 218 | |||
AK152932.1 | - | 3 | 192 | |||
BC051205.1 | - | 2 | 187 | |||
BC054792.1 | - | 2 | 178 | 6832855 | ||
BC055697.1 | - | 2 | 173 | 6411145 | ||
BC076563.1 | - | 3 | 183 | 6411321 | ||
BC081428.1 | - | 2 | 173 | 6412149 | ||
BC086885.1 | - | 3 | 191 | 6763031 | ||
BC087867.1 | - | 3 | 194 | 6543067 |