Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030036.1 | NP_084312.1 | 196 | |||
GenBank mRNA | AK019538.1 | - | 197 |
Gene Symbol: 4921511M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201358.1 | NP_001188287.1 | 196 |
Gene Symbol: Gm10057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243016.1 | NP_001229945.1 | 198 |
Gene Symbol: Gm15140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243017.1 | NP_001229946.1 | 196 |