Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033492.2 | NP_001028664.2 | 1010 | |||
GenBank mRNA | AK144342.1 | - | 1011 | |||
AK144350.1 | - | 976 | ||||
AK165797.1 | - | 994 | ||||
BC150912.1 | - | 955 | 9087823 | |||
BC172073.1 | - | 955 | 9054359 |