Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160137.1 | NP_001153609.1 | 1 | 963 | ||
GenBank mRNA | L04851.1 | - | 1 | 963 |
Gene Symbol: Gm20738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207162.2 | NP_997045.1 | 2 | 1184 | ||
GenBank mRNA | BC049586.1 | - | 2 | 1184 | 6774561 |
Gene Symbol: Gm20747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025241.3 | NP_001020412.1 | 2 | 1091 | ||
GenBank mRNA | BC089498.1 | - | 2 | 1091 | 6744023 | |
L04848.1 | - | 963 |
Gene Symbol: Gm20806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160135.2 | NP_001153607.2 | 1 | 963 | ||
GenBank mRNA | L04849.1 | - | 1 | 963 |
Gene Symbol: Gm20815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017394.2 | NP_001017394.2 | 1192 |
Gene Symbol: Gm20816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160144.1 | NP_001153616.1 | 2 | 1193 | ||
GenBank mRNA | BC100413.1 | - | 2 | 1193 | 30919580 |
Gene Symbol: Gm20823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160143.1 | NP_001153615.1 | 2 | 1193 | ||
GenBank mRNA | BC089467.1 | - | 2 | 1193 | 6772630 |
Gene Symbol: Gm20852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160129.1 | NP_001153601.1 | 2 | 1123 | ||
GenBank mRNA | BC087891.1 | - | 2 | 1123 | 6774426 |
Gene Symbol: Gm20854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160131.1 | NP_001153603.1 | 1 | 965 | ||
GenBank mRNA | L04847.1 | - | 1 | 965 | ||
M38247.1 | - | 1 | 961 |
Gene Symbol: Gm20867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160142.1 | NP_001153614.1 | 2 | 1192 | ||
GenBank mRNA | BC089466.1 | - | 2 | 1192 | 6742357 |
Gene Symbol: Gm20868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006494.1 | - | 2 | 1178 |
Gene Symbol: Gm20917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160136.1 | NP_001153608.1 | 1 | 963 | ||
GenBank mRNA | L04850.1 | - | 1 | 963 |
Gene Symbol: Gm21394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC100414.1 | - | 2 | 1187 | 30920635 |
Gene Symbol: Gm21943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017393.3 | NP_001017393.1 | 2 | 1173 | ||
GenBank mRNA | BC089465.1 | - | 2 | 1173 | 6707203 |
Gene Symbol: Ssty2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023546.3 | NP_076035.3 | 2 | 1180 | ||
GenBank mRNA | L04852.1 | - | 2 | 1179 |