Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195265.1 | NP_001182194.1 | 7 | 1278 | ||
GenBank mRNA | KF041446.1 | - | 7 | 1149 | ||
KF041447.1 | - | 7 | 1149 | |||
KF041448.1 | - | 5 | 1029 | |||
KF041449.1 | - | 6 | 1088 | |||
KF041450.1 | - | 6 | 1088 |